missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human POP4 (aa 3-68) Control Fragment Recombinant Protein

Catalog No. RP109757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109757 Supplier Invitrogen™ Supplier No. RP109757

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144918. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

POP4 is a part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends. It may function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.

Specifications

Accession Number O95707
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 10775
Name Human POP4 (aa 3-68) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1110023P21Rik; hPOP4; hypothetical protein LOC553787; POP4; POP4 (processing of precursor , S. cerevisiae) homolog; POP4 homolog, ribonuclease P/MRP subunit; POP4 protein; processing of precursor 4, ribonuclease P/MRP family, (S. cerevisiae); processing of precursor 4, ribonuclease P/MRP subunit; processing of precursor 4, ribonuclease P/MRP subunit (S. cerevisiae); ribonuclease P protein subunit p29; RPP29; zgc:110597
Common Name POP4
Gene Symbol POP4
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SVIYHALSQKEANDSDVQPSGAQRAEAFVRAFLKRSTPRMSPQAREDQLQRKAVVLEYFTRHKRKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less