missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human PlGF-1 Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $4890.00
Specifications
| Accession Number | P49763 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 5228 |
| Molecular Weight (g/mol) | 29.7 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
10006100UG
|
Gibco™
10006100UG |
100 μg |
Each for $931.06
|
|
|||||
100061MG
|
Gibco™
LS100061MG |
1 mg |
Each for $4,890.00
|
|
|||||
10006250UG
|
Gibco™
10006250UG |
250 μg |
Each for $1,628.00
|
|
|||||
1000625UG
|
Gibco™
LS1000625UG |
25 μg |
Each for $318.00
|
|
|||||
10006500UG
|
Gibco™
10006500UG |
500 μg |
Each for $2,795.00
|
|
|||||
100065UG
|
Gibco™
LS100065UG |
5 μg |
Each for $132.00
|
|
|||||
Description
Recombinant Human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 132 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration.Specifications
| P49763 | |
| protein with no preservative | |
| 29.7 kDa | |
| E. coli | |
| <1 EU/ μg | |
| PlGF-1 | |
| Determined by its ability to chemoattract human monocytes of using a concentration range of 1.0-10.0 ng/ml | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 5228 | |
| Human PlGF-1 | |
| RUO | |
| Placenta Growth Factor; Placenta Growth Factor-1 | |
| PGF | |
| Unconjugated | |
| MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR | |
| E. coli | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |