missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PHF16 (aa 567-690) Control Fragment Recombinant Protein

Catalog No. RP91997
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91997 Supplier Invitrogen™ Supplier No. RP91997

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52240 (PA5-52240. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.

Specifications

Accession Number Q92613
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9767
Name Human PHF16 (aa 567-690) Control Fragment
Quantity 100 μL
Gene Alias 5730598B06; AI851988; AW544806; jade family PHD finger 3; jade family PHD finger protein 3; JADE3; JADE-3; KIAA0215; mKIAA0215; PHD finger protein 16; PHF16; Protein Jade-3
Common Name PHF16
Gene Symbol JADE3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PDSSSSVHSIRNMQVPQESLEMRTKSYPRYPLESKNNRLLASLSHSRSEAKESSPAWRTPSSECYHGQSLGKPLVLQAALHGQSSIGNGKSQPNSKFAKSNGLEGSWSGNVTQKDSSSEMFCDQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less