missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Pellino 2 (aa 297-321) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62754 (PA5-62754. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
E3 ubiquitin ligase catalyzing the covalent attachment of ubiquitin moieties onto substrate proteins. Involved in the TLR and IL-1 signaling pathways via interaction with the complex containing IRAK kinases and TRAF6. Mediates IL1B-induced IRAK1 'Lys-63'-linked polyubiquitination and possibly 'Lys-48'-linked ubiquitination. May be important for LPS- and IL1B-induced MAP3K7-dependent, but not MAP3K3-dependent, NF-kappa-B activation. Can activate the MAP (mitogen activated protein) kinase pathway leading to activation of ELK1. [UniProt]
Specifications
Specifications
| Accession Number | Q9HAT8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57161 |
| Name | Human Pellino 2 (aa 297-321) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AW047589; BB129927; E3 ubiquitin-protein ligase pellino homolog 2; Peli2; pellino 2; pellino E3 ubiquitin protein ligase family member 2; pellino homolog 2; pellino-2; protein pellino homolog 2; RGD1565884; RING-type E3 ubiquitin transferase pellino homolog 2 |
| Common Name | Pellino 2 |
| Show More |