missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PDS5A (aa 1271-1336) Control Fragment Recombinant Protein

Catalog No. RP96638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96638 Supplier Invitrogen™ Supplier No. RP96638

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57756 (PA5-57756. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. NOL5A is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.Nop56p is a yeast nucleolar protein that is part of a complex with the nucleolar proteins Nop58p and fibrillarin. Nop56p is required for assembly of the 60S ribosomal subunit and is involved in pre-rRNA processing. The protein encoded by this gene is similar in sequence to Nop56p and is also found in the nucleolus. Multiple transcript variants encoding several different isoforms have been found for this gene, but the full-length nature of most of them have not been determined.

Specifications

Accession Number Q29RF7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23244
Name Human PDS5A (aa 1271-1336) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9030416H16Rik; Cell proliferation-inducing gene 54 protein; E230024D05Rik; KIAA0648; mKIAA0648; PDS5; PDS5 cohesin associated factor A; PDS5, regulator of cohesion maintenance, homolog A; PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae); PDS5A; PIG54; RGD1307094; SCC112; SCC-112; Sister chromatid cohesion protein 112; sister chromatid cohesion protein PDS5 homolog A
Common Name PDS5A
Gene Symbol PDS5A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GRRPKSESQGNATKNDDLNKPINKGRKRAAVGQESPGGLEAGNAKAPKLQDLAKKAAPAERQIDLQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less