missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PCDHAC1 (aa 705-812) Control Fragment Recombinant Protein

Catalog No. RP106415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106415 Supplier Invitrogen™ Supplier No. RP106415

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65739 (PA5-65739. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences.

Specifications

Accession Number Q9H158
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56135
Name Human PCDHAC1 (aa 705-812) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cadherin-related neuronal receptor c1; CNRc1; PCDHAC1; PCDH-ALPHA-C1; protocadherin alpha C1; protocadherin alpha subfamily C, 1; protocadherin alpha-C1; rCNRvc1
Common Name PCDHAC1
Gene Symbol PCDHAC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TKLHQSPGCCAQSCCRSTEDLRYGSKMVSNPCMTSATIDVTTVERLSQTYLYRASLGLGSDNNSLLLRGEYNAADLRNLATGVGLNLPISCIQIRNRKGDHANVNAMP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less