missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PCDH10 (aa 955-1025) Control Fragment Recombinant Protein

Catalog No. RP107931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107931 Supplier Invitrogen™ Supplier No. RP107931

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67292 (PA5-67292. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene belongs to the protocadherin gene family, a subfamily of the cadherin superfamily. The mRNA encodes a cadherin-related neuronal receptor thought to play a role in the establishment and function of specific cell-cell connections in the brain. This family member contains 6 extracellular cadherin domains, a transmembrane domain and a cytoplasmic tail differing from those of the classical cadherins. Alternatively spliced transcripts encode isoforms with unique cytoplasmic domains.

Specifications

Accession Number Q9P2E7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57575
Name Human PCDH10 (aa 955-1025) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6430521D13Rik; 6430703F07Rik; KIAA1400; LOW QUALITY PROTEIN: protocadherin-10; mKIAA1400; OL-pc; OL-PCDH; OL-protocadherin; ortholog of OL-pcdh; PCDH10; PCDH19; protocadherin 10; protocadherin-10; RGD1565811
Common Name PCDH10
Gene Symbol PCDH10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPSFVPSDGRQAADYRSNLHVPGMDSVPDTEVFETPEAQPGAERSFSTFGKEKALHSTLERKELDGLLTNT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less