missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PBDC1 (aa 48-193) Control Fragment Recombinant Protein

Catalog No. RP92192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92192 Supplier Invitrogen™ Supplier No. RP92192

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51842 (PA5-51842. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PBDC1 is a protein coding gene.

Specifications

Accession Number Q9BVG4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51260
Name Human PBDC1 (aa 48-193) Control Fragment
Quantity 100 μL
Gene Alias 2610029G23Rik; CXorf26; MGC874; Pbdc1; polysaccharide biosynthesis domain containing 1; polysaccharide biosynthesis domain-containing protein 1; Protein PBDC1; UPF0368 protein Cxorf26; UPF0368 protein Cxorf26 homolog
Common Name PBDC1
Gene Symbol Pbdc1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EVYYKLISSVDPQFLKLTKVDDQIYSEFRKNFETLRIDVLDPEELKSESAKEKWRPFCLKFNGIVEDFNYGTLLRLDCSQGYTEENTIFAPRIQFFAIEIARNREGYNKAVYISVQDKEGEKGVNNGGEKRADSGEEENTKNGGEK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less