missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAQR9 (aa 257-277) Control Fragment Recombinant Protein

Catalog No. RP101587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP101587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP101587 Supplier Invitrogen™ Supplier No. RP101587

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62690. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q6ZVX9
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 344838
Name Human PAQR9 (aa 257-277) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Membrane progesterone P4 receptor epsilon; Membrane progesterone receptor epsilon; Membrane progestin receptor epsilon; mPR epsilon; PAQR9; Progesterone and adipoQ receptor family member 9; Progestin and adipoQ receptor family member 9; progestin and adipoQ receptor family member IX
Common Name PAQR9
Gene Symbol PAQR9
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IMLESWLFDLRGENPTLFVHF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less