missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OXER1 (aa 360-409) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP101422

Catalog No. RP101422


Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (33%), Rat (33%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TG1019 is a G protein-coupled receptor that functions as a chemoattractant for eosinophils and neutrophils. TG1019 is expressed in various tissues except the brain, with the highest expression in liver, kidney, peripheral leukocyte, lung and spleen. TG1019 interacts with the protein OXE to form TG1019/OXE, a Galpha i/o protein-coupled receptor. Signals from TG1019 are transduced via a Galpha i/o protein to PLC/calcium, MEK/ERK and PI3K/Akt pathways. Signal transduction from TG1019 following stimulation with 5-oxo-6E,8Z,11Z,14Z eicosatetraenoic acid (5-oxo-ETE) induces intracellular calcium mobilization and rapid activation of MEK/ERK and PI3K/Akt pathways. TG1019 also may play a role in downregulating cAMP production.
Specifications

Specifications

Q8TDS5
Blocking Assay, Control
165140
100 μL
5-oxo-ETE acid G-protein-coupled receptor 1; 5-oxo-ETE G-protein coupled receptor; GPCR; GPR170; G-protein coupled receptor 170; G-protein coupled receptor R527; G-protein coupled receptor TG1019; HEOAD54; OXE receptor; OXER1; oxoeicosanoid (OXE) receptor 1; oxoeicosanoid receptor 1; TG1019
OXER1
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human OXER1 (aa 360-409) Control Fragment
RUO
OXER1
Unconjugated
Recombinant
PNFLHQSRALLGLTRGRQGPVSDESSYQPSRQWRYREASRKAEAIGKLKV
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions