missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OR7C2 (aa 293-319) Control Fragment Recombinant Protein

Catalog No. RP100790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100790 Supplier Invitrogen™ Supplier No. RP100790
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (48%), Rat (48%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61487 (PA5-61487. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.

Specifications

Accession Number O60412
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 26658
Name Human OR7C2 (aa 293-319) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CIT-HSP-87M17; Olfactory receptor 19-18; olfactory receptor 7C2; Olfactory receptor 7C3; olfactory receptor family 7 subfamily C member 2; olfactory receptor OR19-22; olfactory receptor, family 7, subfamily C, member 2; olfactory receptor, family 7, subfamily C, member 3; OR19-18; OR7C2; OR7C3
Common Name OR7C2
Gene Symbol OR7C2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NKDMKGSLGRLLLRATSLKEGTIAKLS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less