missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human OR51E1 (aa 295-317) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101516
Description
Highest antigen sequence indentity to the following orthologs: Mouse (78%), Rat (78%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Odorant receptor (Potential).Specifications
| Q8TCB6 | |
| Blocking Assay, Control | |
| 143503 | |
| 100 μL | |
| DGPCR; D-GPCR; Dresden-G-protein-coupled receptor; GPR136; GPR164; G-protein coupled receptor 164; Olfactory receptor 51E1; olfactory receptor 52A3; olfactory receptor family 51 subfamily E member 1; olfactory receptor OR11-15; olfactory receptor, family 51, subfamily E, member 1 pseudogene; olfactory receptor, family 52, subfamily A, member 3 pseudogene; OR51E1; OR51E1P; OR52A3P; POGR; prostate overexpressed G protein-coupled receptor; prostate-overexpressed G protein-coupled receptor; Prostate-specific G protein-coupled receptor 2; PSGR2 | |
| OR51E1 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human OR51E1 (aa 295-317) Control Fragment | |
| RUO | |
| OR51E1 | |
| Unconjugated | |
| Recombinant | |
| VKTKEIRQRILRLFHVATHASEP | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |