missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OR4M2 (aa 172-198) Control Fragment Recombinant Protein

Catalog No. RP104522
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104522 Supplier Invitrogen™ Supplier No. RP104522
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63624 (PA5-63624. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

OR4M2 is an odorant receptor (Potential).

Specifications

Accession Number Q8NGB6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 390538
Name Human OR4M2 (aa 172-198) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Olfactory receptor 4M2; olfactory receptor family 4 subfamily M member 2; olfactory receptor OR15-3; olfactory receptor, family 4, subfamily M, member 2; OR15-3; OR4M2
Common Name OR4M2
Gene Symbol OR4M2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NELDSYFCDITQVVRIACANTFPEELV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less