missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OR2B11 (aa 3-23) Control Fragment Recombinant Protein

Catalog No. RP100608
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100608 Supplier Invitrogen™ Supplier No. RP100608

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61525 (PA5-61525. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Odorant receptor.

Specifications

Accession Number Q5JQS5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 127623
Name Human OR2B11 (aa 3-23) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor 2B11; olfactory receptor family 2 subfamily B member 11; olfactory receptor, family 2, subfamily B, member 11; OR2B11
Common Name OR2B11
Gene Symbol OR2B11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SDNHSFLGDSPKAFILLGVSD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less