missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human OR10Q1 Control Fragment Recombinant Protein

Catalog No. RP100610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100610 Supplier Invitrogen™ Supplier No. RP100610

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61385 (PA5-61385. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Odorant receptor.

Specifications

Accession Number Q8NGQ4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 219960
Name Human OR10Q1 Control Fragment
Quantity 100 μL
Gene Alias olfactory receptor 10Q1; olfactory receptor family 10 subfamily Q member 1; olfactory receptor OR11-233; olfactory receptor, family 10, subfamily Q, member 1; OR10Q1; OR11-233
Common Name OR10Q1
Gene Symbol OR10Q1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GKLVFNQSDPTEFVFRAFTTATEFQV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less