missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NYAP2 (aa 364-436) Control Fragment Recombinant Protein

Catalog No. RP99644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99644 Supplier Invitrogen™ Supplier No. RP99644
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61026 (PA5-61026. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.

Specifications

Accession Number Q9P242
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57624
Name Human NYAP2 (aa 364-436) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9430031J16Rik; Jr6; Kiaa1486; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2; NEWGENE_1305560; NYAP2
Common Name NYAP2
Gene Symbol NYAP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PVLENVSYMKQPAGASPSTLPSHVPGHAKLEKEQAAALGPASATPALSSSPPPPSTLYRTQSPHGYPKSHSTS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less