missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NUP93 (aa 29-144) Control Fragment Recombinant Protein

Catalog No. RP91837
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91837 Supplier Invitrogen™ Supplier No. RP91837
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53674 (PA5-53674. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. This gene encodes a nucleoporin protein that localizes both to the basket of the pore and to the nuclear entry of the central gated channel of the pore. The encoded protein is a target of caspase cysteine proteases that play a central role in programmed cell death by apoptosis. Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Accession Number Q8N1F7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9688
Name Human NUP93 (aa 29-144) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410008G02Rik; 93 kDa nucleoporin; Allele: hi4; CBP-interacting protein 4; Cip4; dead eye; dead eye protein; dye; Kiaa0095; NIC96; NPHS12; nuclear pore complex protein Nup93; nucleoporin 93; nucleoporin 93 kDa; nucleoporin Nup93; nup93
Common Name NUP93
Gene Symbol Nup93
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VERNLQEIQQAGERLRSRTLTRTSQETADVKASVLLGSRGLDISHISQRLESLSAATTFEPLEPVKDTDIQGFLKNEKDNALLSAIEESRKRTFGMAEEYHRESMLVEWEQVKQRI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less