missing translation for 'onlineSavingsMsg'
Learn More

Human NTAL Alexa Fluor 750-conjugated Antibody, R&D Systems™

Catalog No. IC4066S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4066S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC4066S100 Supplier Bio-Techne Supplier No. IC4066S100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

NTAL Monoclonal specifically detects NTAL in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen NTAL
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 440005
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9GZY6
Gene Alias LAB, LABWilliams-Beuren syndrome chromosomal region 15 protein, Linker for activation of B-cells, linker for activation of T cells family, member 2, linker for activation of T-cells family member 2, Membrane-associated adapter molecule, Non-T-cell activation linker, NTALWilliams-Beuren syndrome chromosomal region 5 protein, WBS15, WBSCR15, WBSCR5, WBSCR5HSPC046, Williams-Beuren syndrome chromosome region 5, WSCR5
Gene Symbols LAT2
Host Species Mouse
Immunogen E. coli-derived recombinant human NTAL Met1-Ala243 Accession # Q9GZY6
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7462
Test Specificity Detects human NTAL in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)