missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NT5M (aa 103-166) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP100000

Catalog No. RP100000

Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60504 (PA5-60504. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Dephosphorylates specifically the 5' and 2'(3')- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.
Specifications

Specifications

Q9NPB1
Blocking Assay, Control
56953
100 μL
2010013E09Rik; 5' nucleotidase, mitochondrial; 5(3)-deoxyribonucleotidase; 5'(3')-deoxyribonucleotidase, mitochondrial; 5',3'-deoxyribonucleotidase, mitochondrial; 5',3'-nucleotidase, mitochondrial; AI846937; Deoxy-5'-nucleotidase 2; DNT2; dNT-2; mdN; mitochondrial 5' nucleotidase; NT5M
NT5M
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human NT5M (aa 103-166) Control Fragment
RUO
NT5M
Unconjugated
Recombinant
FELEPLPGAVEAVKEMASLQNTDVFICTSPIKMFKYCPYEKYAWVEKYFGPDFLEQIVLTRDKT
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions