missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NRBP2 (aa 366-493) Control Fragment Recombinant Protein

Catalog No. RP104595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104595 Supplier Invitrogen™ Supplier No. RP104595

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65039 (PA5-65039. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.

Specifications

Accession Number Q9NSY0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 340371
Name Human NRBP2 (aa 366-493) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC011468; NRBP2; nuclear receptor binding protein 2; nuclear receptor-binding protein 2; pp9320; Transformation-related gene 16 protein; transformation-related protein 16; TRG16; TRG-16
Common Name NRBP2
Gene Symbol Nrbp2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MELDKFLEDVRNGIYPLMNFAATRPLGLPRVLAPPPEEVQKAKTPTPEPFDSETRKVIQMQCNLERSEDKARWHLTLLLVLEDRLHRQLTYDLLPTDSAQDLASELVHYGFLHEDDRMKLAAFLESTF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less