missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NRBP2 (aa 259-310) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP96533
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56048 (PA5-56048. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.Specifications
| Q9NSY0 | |
| Blocking Assay, Control | |
| 340371 | |
| 100 μL | |
| BC011468; NRBP2; nuclear receptor binding protein 2; nuclear receptor-binding protein 2; pp9320; Transformation-related gene 16 protein; transformation-related protein 16; TRG16; TRG-16 | |
| Nrbp2 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human NRBP2 (aa 259-310) Control Fragment | |
| RUO | |
| NRBP2 | |
| Unconjugated | |
| Recombinant | |
| DTRVTEEAIARARHSLSDPNMREFILCCLARDPARRPSAHSLLFHRVLFEVH | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |