missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2 A(17); neuropilin-2 A(22); neuropilin-2 b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2 |
|---|---|
| Assay Range | 8.1 to 2,000 ng/mL |
| Assay Sensitivity | 8.1 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH336RBX5
|
Invitrogen™
EH336RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH336RB
|
Invitrogen™
EH336RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Neuropilin-2 quantitates human Neuropilin-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Neuropilin-2.
This gene encodes a member of the neuropilin family of receptor proteins. The encoded transmembrane protein binds to SEMA3C protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C) and SEMA3F protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F), and interacts with vascular endothelial growth factor (VEGF). This protein may play a role in cardiovascular development, axon guidance, and tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| 1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2 A(17); neuropilin-2 A(22); neuropilin-2 b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2 | |
| 8.1 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| NRP2 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Neuropilin 2 | |
| 4 hrs 45 mins |
| 8.1 to 2,000 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 8828 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |