missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NEURL4 (aa 356-448) Control Fragment Recombinant Protein

Catalog No. RP100345
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100345 Supplier Invitrogen™ Supplier No. RP100345

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63108 (PA5-63108. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q96JN8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84461
Name Human NEURL4 (aa 356-448) Control Fragment
Quantity 100 μL
Gene Alias KIAA1787; neuralized E3 ubiquitin protein ligase 4; neuralized homolog 4; neuralized-like protein 4; NEURL4
Common Name NEURL4
Gene Symbol NEURL4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PLRDNEMFEIRIDKLVDKWSGSIEIGVTTHNPNSLEYPATMTNLQSGTIMMSGCGILTNGKGTRREYCEFSLDELQEGDHIGLTRKSNSALHF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less