missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human NETO2 (NP_060562.3) VersaClone cDNA

Specifications
Specifications
| Gene | NETO2 |
| Accession Number | NP_060562.3 |
| Gene ID (Entrez) | 81831 |
| Gene Alias | Brain-specific transmembrane protein containing 2 CUB and 1 LDL-receptor classA domains protein 2, BTCL2, FLJ10430, FLJ90456, NEOT2, NEOT2FLJ14724, neuropilin (NRP) and tolloid (TLL)-like 2, neuropilin and tolloid-like protein 2 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |