missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; nestin |
|---|---|
| Assay Range | 0.32 to 75 ng/mL |
| Assay Sensitivity | 0.32 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH334RBX5
|
Invitrogen™
EH334RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH334RB
|
Invitrogen™
EH334RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Nestin/MMEL1 quantitates human Nestin/MMEL1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nestin/MMEL1.
Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.Specifications
| AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; nestin | |
| 0.32 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| Nes | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Nestin | |
| 4 hrs 45 mins |
| 0.32 to 75 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 10763 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |