missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | AI607786; Calnuc; DNA-binding protein NEFA; epididymis secretory protein Li 109; Gastric cancer antigen Zg4; HEL-S-109; NEFA; nesfatin 1; nesfatin-1; Nucb2; nucleobindin 2; nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobinding 2; p54; prepronefastin; prepronesfatin |
|---|---|
| Assay Range | 0.25 to 60 ng/mL |
| Assay Sensitivity | 0.25 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH333RBX5
|
Invitrogen™
EH333RBX5 |
5 x 96 Tests |
N/A
|
|
|||||
EH333RB
|
Invitrogen™
EH333RB |
96 Tests |
N/A
|
|
|||||
Description
Human Nesfatin-1 quantitates human Nesfatin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Nesfatin-1.
NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.Specifications
| AI607786; Calnuc; DNA-binding protein NEFA; epididymis secretory protein Li 109; Gastric cancer antigen Zg4; HEL-S-109; NEFA; nesfatin 1; nesfatin-1; Nucb2; nucleobindin 2; nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobinding 2; p54; prepronefastin; prepronesfatin | |
| 0.25 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| NUCB2 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| NUCB2 | |
| 4 hrs 45 mins |
| 0.25 to 60 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4925 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |