missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | B230343H07Rik; IDH3GL; l7R6; Nell1; NEL-like 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; neural EGFL like 1; neural epidermal growth factor-like 1; NRP1; Protein kinase C-binding protein NELL1 |
|---|---|
| Assay Range | 10 to 200 ng/mL |
| Assay Sensitivity | 10 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH332RBX5
|
Invitrogen™
EH332RBX5 |
5 x 96 Tests |
N/A
|
|
|||||
EH332RB
|
Invitrogen™
EH332RB |
96 Tests |
N/A
|
|
|||||
Description
Human NELL-1 quantitates human NELL-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human NELL-1.
This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| B230343H07Rik; IDH3GL; l7R6; Nell1; NEL-like 1; NEL-like 1 (chicken); NEL-like protein 1; Nel-related protein 1; neural EGFL like 1; neural epidermal growth factor-like 1; NRP1; Protein kinase C-binding protein NELL1 | |
| 10 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| NELL1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| NELL1 | |
| 4 hrs 45 mins |
| 10 to 200 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4745 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |