missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NDUFA11 (aa 42-62) Control Fragment Recombinant Protein

Catalog No. RP105555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105555 Supplier Invitrogen™ Supplier No. RP105555
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (62%), Rat (62%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67228 (PA5-67228. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NDUFA11 encodes a subunit of the membrane-bound mitochondrial complex I. Complex I is composed of numerous subunits and functions as the NADH-ubiquinol reductase of the mitochondrial electron transport chain. Mutations in this gene are associated with severe mitochondrial complex I deficiency. Alternate splicing results in multiple transcript variants.

Specifications

Accession Number Q86Y39
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 126328
Name Human NDUFA11 (aa 42-62) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2010012C24Rik; AV006275; B14.7; CI-B14.7; complex I B14.7 subunit; complex I-B14.7; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 11, 14.7 kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 11; NADH:ubiquinone oxidoreductase subunit A11; NADH-ubiquinone oxidoreductase subunit B14.7; NDUFA11
Common Name NDUFA11
Gene Symbol NDUFA11
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TLNPPGTFLEGVAKVGQYTFT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less