missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NAGS (aa 287-395) Control Fragment Recombinant Protein

Catalog No. RP105775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105775 Supplier Invitrogen™ Supplier No. RP105775

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65010. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

NAGS is a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.The N-acetylglutamate synthase gene encodes a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. This gene may regulate ureagenesis by altering NAG availability and, thereby, CPSI activity. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.

Specifications

Accession Number Q8N159
Concentration 2.01 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 162417
Name Human NAGS (aa 287-395) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1700120E20Rik; AGAS; Amino-acid acetyltransferase; amino-acid N-acetyltransferase; ARGA; LOW QUALITY PROTEIN: N-acetylglutamate synthase, mitochondrial; MGC133025; N-acetylglutamate synthase; N-acetylglutamate synthase conserved domain form; N-acetylglutamate synthase long form; N-acetylglutamate synthase short form; N-acetylglutamate synthase, conserved domain form; N-acetylglutamate synthase, mature form; N-acetylglutamate synthase, mitochondrial; Nags; NAGS-C; NAGS-M; RGD1565783
Common Name NAGS
Gene Symbol NAGS
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PTKIIFLNNTGGLRDSSHKVLSNVNLPADLDLVCNAEWVSTKERQQMRLIVDVLSRLPHHSSAVITAASTLLTELFSNKGSGTLFKNAERMLRVRSLDKLDQGRLVDLV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less