missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MYL4 (aa 29-54) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101602
Description
Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84091 (PA5-84091. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Regulatory light chain of myosin. Does not bind calcium.Specifications
| P12829 | |
| Blocking Assay, Control | |
| 4635 | |
| 100 μL | |
| ALC1; AMLC; ELC; ELC1a; GT1; MLC1; MLC1a; MLC1EMB; MYL4; Myla; myosin light chain 1, atrial isoform; myosin light chain 1, atrial/fetal isoform; myosin light chain 1, embryonic muscle/atrial isoform; Myosin light chain 4; myosin light chain alkali GT-1 isoform; myosin light chain, alkali, cardiac atria; myosin, atrial/fetal muscle, light chain; myosin, light chain 4; myosin, light chain 4, alkali; atrial, embryonic; myosin, light polypeptide 4; myosin, light polypeptide 4, alkali; atrial, embryonic; PRO1957 | |
| MYL4 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human MYL4 (aa 29-54) Control Fragment | |
| RUO | |
| MYL4 | |
| Unconjugated | |
| Recombinant | |
| PAPEAPKEPAFDPKSVKIDFTADQIE | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |