missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MTFR1 (aa 261-332) Control Fragment Recombinant Protein

Catalog No. RP93736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93736 Supplier Invitrogen™ Supplier No. RP93736
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54709 (PA5-54709. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.

Specifications

Accession Number Q15390
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9650
Name Human MTFR1 (aa 261-332) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1300002C08Rik; 1700080D04Rik; 2810026O10Rik; 4930579E05Rik; chondrocyte protein with a poly-proline region; CHPPR; FAM54A2; Kiaa0009; Mitochondrial fission regulator 1; Mtfr1; RGD1304936
Common Name MTFR1
Gene Symbol Mtfr1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KPKPVDATDPAALIAEALKKKFAYRYRSDSQDEVEKGIPKSESEATSERVLFGPHMLKPTGKMKALIENVSD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less