missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MRPL39 (aa 249-332) Control Fragment Recombinant Protein

Catalog No. RP91811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91811 Supplier Invitrogen™ Supplier No. RP91811

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53785. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Two transcript variants encoding distinct isoforms have been described. A pseudogene corresponding to this gene is found on chromosome 5q.

Specifications

Accession Number Q9NYK5
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 54148
Name Human MRPL39 (aa 249-332) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 39S ribosomal protein L39, mitochondrial; 39S ribosomal protein L5, mitochondrial; C21orf8; C21orf92; L39mt; L5mt; Mitochondrial large ribosomal subunit protein mL39; mitochondrial ribosomal protein L39; MRPL39; MRP-L39; MRPL5; MRP-L5; MSTP003; ORF22; PRED22; PRED66; ribosomal protein, mitochondrial, L5; RPML5
Common Name MRPL39
Gene Symbol MRPL39
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ERIVKLHRIGDFIDVSEGPLIPRTSICFQYEVSAVHNLQPTQPSLIRRFQGVSLPVHLRAHFTIWDKLLERSRKMVTEDQSKAT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less