missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse SOX2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC20181G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC20181G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC20181G100 Supplier Bio-Techne Supplier No. IC20181G100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse MonoclonalAntibody

SOX2 Monoclonal specifically detects SOX2 in Human∣Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen SOX2
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 245610R
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P48431
Gene Alias ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2
Host Species Mouse
Immunogen E.coli-derived recombinant human SOX2 Gly135-Met317 Accession # P48431
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6657
Test Specificity Detects human and mouse SOX2 in Western blots.
Target Species Human, Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2a
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)