missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human/Mouse/Rat Thymosin-beta4 Recombinant Protein, PeproTech®
Recombinant Protein
$125.66 - $5780.79
Specifications
| Accession Number | P20065, P62328, P62329 |
|---|---|
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 19241, 7114, 81814 |
| Name | Human/Mouse/Rat Thymosin-beta4 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
14014100UG
|
Gibco™
14014100UG |
100 μg |
Each for $302.74
|
|
|||||
140141MG
|
Gibco™
140141MG |
2 x 500 μg |
Each for $1,700.00
|
|
|||||
|
14-014-01M
|
Gibco™
1401401M |
2 x 500 μg |
Each for $5,780.79
|
|
|||||
1401420UG
|
Gibco™
1401420UG |
20 μg |
Each for $125.66
|
|
|||||
14014250UG
|
Gibco™
14014250UG |
250 μg |
Each for $596.00
|
|
|||||
14014500UG
|
Gibco™
14014500UG |
500 μg |
Each for $1,024.00
|
|
|||||
Description
140-14-1MG will be provided as 2 x 500 μg (140-14-500UG). Recombinant Human/Murine/Rat Thymosin-beta4 is a 5.2 kDa glycoprotein containing 45 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Thymosin-beta4 is a small, actin-sequestering protein belonging to the thymosin-beta family that is found at high concentrations within the spleen, thymus, and peritoneal macrophages, where it is most notably responsible for the organization of cytoskeletal structure. In mammalian tissues, this protein acts as a modulator for the polymerization/depolymerization of actin through the formation of a 1:1 complex with the monomer G (globular)-actin, and inhibits actin's polymerization to form F (filamentous) actin, which together with other proteins binds microfilaments to construct the cytoskeleton. Commonly found at significant quantities within the brain, lungs, liver, kidneys, testes, and heart, thymosin-beta4 has also been shown to be synthesized by cells unrelated to the reticuloendothelial system, such as myoblasts and fibroblasts, and expressed at irregular levels by several hemopoietic cell lines, malignant lymphoid cells and myeloma cells. In addition to regulating actin polymerization, research has also found thymosin-beta4 to stimulate the secretion of hypothalamic luteinizing hormone-releasing hormone and luteinizing hormone, inhibit the migration of peritoneal macrophages, induce phenotypic changes in T cell lines during early host defense mechanisms, and inhibit the progression of hematopoietic pluripotent stem cells into the S-phase.Specifications
| P20065, P62328, P62329 | |
| protein with no preservative | |
| Human/Mouse/Rat Thymosin-beta4 | |
| RUO | |
| TMSB4X | |
| Recombinant | |
| Lyophilized |
| Functional Assay | |
| 19241, 7114, 81814 | |
| E. coli | |
| Fx; Hematopoietic system regulatory peptide; prothymosin beta 4; prothymosin beta-4; Ptmb4; Seraspenide; T beta 4; t beta-4; Tb4; TB4X; TB4Xbeta 4; Thyb4; thymosin; thymosin beta 4; thymosin beta 4, X-linked; thymosin beta-4; thymosin, beta 4, x chromosome; Tmsb4; Tmsb4x | |
| Unconjugated | |
| RMSDKPDMAE IEKFDKSKLK KTETQEKNPL PSKETIEQEK QAGES | |
| ≥ 95% by SDS-PAGE gel and HPLC analyses. |