missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech®
Recombinant Protein
Supplier: Gibco™ LS120111MG
Description
120-11-1MG will be provided as 2 x 500 μg (120-11-500UG). Recombinant Human/Murine/Rat GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8, sharing 90% amino acid sequence identity. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
GDF11 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development.Specifications
| O95390, Q9Z1W4, Q9Z217 | |
| protein with no preservative | |
| 25 | |
| 2 x 500 μg | |
| -20°C | |
| <1 EU/μg | |
| GDF11 | |
| Unconjugated | |
| NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |
| Functional Assay | |
| 10220, 14561, 29454 | |
| Human/Mouse/Rat GDF-11 | |
| E. coli | |
| RUO | |
| BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11 | |
| Human, Mouse, Rat | |
| Recombinant | |
| Lyophilized |