missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse MYF-5 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Catalog No. IC4027S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC4027S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC4027S100 Supplier Bio-Techne Supplier No. IC4027S100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MYF-5 Monoclonal specifically detects MYF-5 in Human∣Mouse samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen MYF-5
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 593128
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P13349
Gene Alias BHLHC2, bHLHc2Class C basic helix-loop-helix protein 2, myf-5, MYF5, myogenic factor 5
Gene Symbols MYF5
Host Species Mouse
Immunogen E. coli-derived recombinant human MYF-5 Met1-Leu255 Accession # P13349
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4617
Test Specificity Detects human MYF-5 in direct ELISAs.
Target Species Human, Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2a
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)