missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MORN1 (aa 61-192) Control Fragment Recombinant Protein

Catalog No. RP88838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP88838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP88838 Supplier Invitrogen™ Supplier No. RP88838

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110700 (PA5-110700. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function of this protein remains unknown.

Specifications

Accession Number Q5T089
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79906
Name Human MORN1 (aa 61-192) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias MORN repeat containing 1; MORN repeat-containing protein 1; MORN1
Common Name MORN1
Gene Symbol MORN1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YYEGAFVDGEITGEGRRHWAWSGDTFSGQFVLGEPQGYGVMEYKAGGCYEGEVSHGMREGHGFLVDRDGQVYQGSFHDNKRHGPGQMLFQNGDKYDGDWVRDRRQGHGVLRCADGSTYKGQWHSDVFSGLGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less