missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MGST3 Control Fragment Recombinant Protein

Catalog No. RP106451
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106451 Supplier Invitrogen™ Supplier No. RP106451

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65773 (PA5-65773. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The MAPEG family consists of six human proteins, several of which are involved the production of leukotrienes and prostaglandin E, important mediators of inflammation. This gene encodes an enzyme which catalyzes the conjugation of leukotriene A4 and reduced glutathione to produce leukotriene C4. This enzyme also demonstrates glutathione-dependent peroxidase activity towards lipid hydroperoxides.

Specifications

Accession Number O14880
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4259
Name Human MGST3 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2010012L10Rik; 2010306B17Rik; 2700004G04Rik; AA516734; Glutathione peroxidase MGST3; GST-III; LTC4 synthase MGST3; MGST3; microsomal glutathione S-transferase; microsomal glutathione S-transferase 3; Microsomal glutathione S-transferase III; microsomal GST-3; Microsomal GST-III
Common Name MGST3
Gene Symbol MGST3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPWRGRRFRLCRRKMAVLSK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less