missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MGAM2 (aa 47-121) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (45%), Rat (45%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63162. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MGAM2 (Maltase-Glucoamylase 2 (Putative)) is a Protein Coding gene. Among its related pathways are Metabolism and Galactose metabolism. Gene Ontology (GO) annotations related to this gene include carbohydrate binding and glucan 1,4-alpha-glucosidase activity. An important paralog of this gene is MGAM. [GeneCards].
Specifications
Specifications
| Accession Number | Q2M2H8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Neutralization, Control |
| Formulation | PBS, 1M urea with no preservative; pH 7.4 |
| Gene ID (Entrez) | 93432 |
| Name | Human MGAM2 (aa 47-121) Control Fragment |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 100 μL |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |