missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human METTL23 (aa 119-190) Control Fragment Recombinant Protein

Catalog No. RP93474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93474 Supplier Invitrogen™ Supplier No. RP93474

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139771. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene functions as a transcription factor regulator in the transcriptional pathway for human cognition. It is a partner of the alpha subunit of the GA-binding protein transcription factor. Mutations in this gene cause mild autosomal recessive intellectual disability. Alternative splicing results in multiple transcript variants.

Specifications

Accession Number Q86XA0
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 124512
Name Human METTL23 (aa 119-190) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C17orf95; methyltransferase like 23; methyltransferase-like protein 23; METTL23; MRT44
Common Name METTL23
Gene Symbol METTL23
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LMHKNPKVQLWSTYQVRSADWSLEALLYKWDMKCVHIPLESFDADKEDIAESTLPGRHTVEMLVISFAKDSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less