missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MESDC2 (aa 40-125) Control Fragment Recombinant Protein

Catalog No. RP104741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104741 Supplier Invitrogen™ Supplier No. RP104741

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111140. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MESDC2 chaperone specifically assisting the folding of beta-propeller/EGF modules within the family of low-density lipoprotein receptors (LDLRs). Acts as a modulator of the Wnt pathway through chaperoning the coreceptors of the canonical Wnt pathway, LRP5 and LRP6, to the plasma membrane. Essential for specification of embryonic polarity and mesoderm induction. Plays an essential role in neuromuscular junction (NMJ) formation by promoting cell-surface expression of LRP4. May regulate phagocytosis of apoptotic retinal pigment epithelium (RPE) cells.

Specifications

Accession Number Q14696
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 23184
Name Human MESDC2 (aa 40-125) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2210015O11Rik; AW537813; BOCA; KIAA0081; LDLR chaperone MESD; LRP chaperone MESD; MESD; Mesdc2; MESDM; mesoderm development candiate 2; mesoderm development candidate 2; Mesoderm development LRP chaperone MESD; Mesoderm development protein; mKIAA0081; msd; renal carcinoma antigen NY-REN-61; UNQ1911/PRO4369
Common Name MESDC2
Gene Symbol Mesd
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TPDESTPPPRKKKKDIRDYNDADMARLLEQWEKDDDIEEGDLPEHKRPSAPVDFSKIDPSKPESILKMTKKGKTLMMFVTVSGSPT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less