missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Meprin alpha ELISA Kit

Catalog No. EH321RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH321RBX5 5 x 96 Tests
EH321RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH321RBX5 Supplier Invitrogen™ Supplier No. EH321RBX5
Only null left

ELISA

Human Meprin alpha quantitates human Meprin alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Meprin alpha.

MEP1A (Meprin A Subunit Alpha) is a Protein Coding gene. Among its related pathways are Collagen chain trimerization. Gene Ontology (GO) annotations related to this gene include metalloendopeptidase activity and metallopeptidase activity.

Specifications

Accession Number AI098089; AW107200; bA268F1.1 (meprin A alpha (PABA peptide hydrolase)); E-24.18; Endopeptidase-2; Endopeptidase-24.18 subunit alpha; Mep1; MEP-1; Mep1a; Mep-1 A; meprin 1 alpha; meprin A alpha; meprin A alpha-subunit; Meprin A subunit alpha; meprin A, alpha (PABA peptide hydrolase); meprin alpha; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; PABA peptide hydrolase; PPH alpha; PPHA
Assay Range 2 to 400 ng/mL
Assay Sensitivity 2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4224
Gene Symbol MEP1A
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target MEP1A
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less