missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | Crtm2; Matn2; matrilin 2; matrilin-2; testis tissue sperm-binding protein Li 94 mP; UNQ193/PRO219 |
|---|---|
| Assay Range | 0.33 to 80 ng/mL |
| Assay Sensitivity | 0.33 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH313RBX5
|
Invitrogen™
EH313RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH313RB
|
Invitrogen™
EH313RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Matrilin-2 quantitates human Matrilin-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Matrilin-2.
Matrilin (MATNs) are a family of non-collagenous extracellular matrix proteins consisting of four known members that have been proposed to play key roles in the formation of both collagen-dependent and collagen-independent filamentous networks. The matrilin family all share a structure made up of von Willebrand factor A domains, epidermal growth factor-like domains and a coiled coil alpha-helical module. MATN1 and MATN3 are expressed mainly in cartilage, while MATN2 and MATN4 occur in a wide variety of extracellular matrices. The matrilin genes are strictly and differently regulated and their expression may serve as markers for cellular differentiation and diseases such as astrocytoma and liver carcinoma. Recent studies show that MATN2 is a permissive substrate for axonal growth and cell migration, and it is required for successful nerve regeneration.Specifications
| Crtm2; Matn2; matrilin 2; matrilin-2; testis tissue sperm-binding protein Li 94 mP; UNQ193/PRO219 | |
| 0.33 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| MATN2 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Matrilin 2 | |
| 4 hrs 45 mins |
| 0.33 to 80 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4147 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |