missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MALRD1 (aa 1784-1846) Control Fragment Recombinant Protein

Catalog No. RP105907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105907 Supplier Invitrogen™ Supplier No. RP105907
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65158. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Enhances production and/or transport of FGF19 and thus has a role in regulation of bile acid synthesis.

Specifications

Accession Number Q5VYJ5
Concentration 3 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 340895
Name Human MALRD1 (aa 1784-1846) Control Fragment
pH Range 7.4
Purification Method Metal-chelate chromatography
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias bA265G8.2; C10orf112; DIET1; MALRD1; MAM and LDL receptor class A domain containing 1; MAM and LDL-receptor class A domain-containing protein 1; MAM and LDL-receptor class A domain-containing protein C10orf112; novel MAM domain containing protein
Common Name MALRD1
Gene Symbol MALRD1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SSGSKEGSVARITTSKSFPASLGMCTVRFWFYMIDPRSMGILKVYTIEESGLNILVWSVIGNK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less