missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | AAT9; MAGP2; MAGP-2; MFAP5; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; microfibrillar associated protein 5; microfibrillar-associated protein 5; MP25; THE1A-MFAP5 |
|---|---|
| Assay Range | 0.04 to 10 ng/mL |
| Assay Sensitivity | 0.04 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH312RBX5
|
Invitrogen™
EH312RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH312RB
|
Invitrogen™
EH312RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human MAGP-2/MFAP5 quantitates human MAGP-2/MFAP5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human MAGP-2/MFAP5.
This gene encodes a 25-kDa microfibril-associated glycoprotein which is rich in serine and threonine residues. It lacks a hydrophobic carboxyl terminus and proline-, glutamine-, and tyrosine-rich regions, which are characteristics of a related 31- kDa microfibril-associated glycoprotein (MFAP2). The close similarity between these two proteins is confined to a central region of 60 aa where precise alignment of 7 cysteine residues occurs. The structural differences suggest that this encoded protein has some functions that are distinct from those of MFAP2.Specifications
| AAT9; MAGP2; MAGP-2; MFAP5; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; microfibrillar associated protein 5; microfibrillar-associated protein 5; MP25; THE1A-MFAP5 | |
| 0.04 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| MFAP5 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| MFAP5 | |
| 4 hrs 45 mins |
| 0.04 to 10 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 8076 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |