missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MACF1 Control Fragment Recombinant Protein

Catalog No. RP107619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107619 Supplier Invitrogen™ Supplier No. RP107619

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (23%), Rat (23%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66976 (PA5-66976. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).

Specifications

Accession Number Q9UPN3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23499
Name Human MACF1 Control Fragment
Quantity 100 μL
Gene Alias 620 kDa actin binding protein; 620 kDa actin-binding protein; ABP620; ACF7; Aclp7; Actin cross-linking family 7; actin cross-linking family protein 7; actin-crosslinking protein 7; FLJ45612; FLJ46776; KIAA0465; KIAA0754; KIAA1251; MACF; MACF1; macrophin 1; macrophin 1 isoform; Macrophin-1; microtubule-actin crosslinking factor 1; Microtubule-actin cross-linking factor 1; Microtubule-actin cross-linking factor 1, isoforms 1/2/3/5; mKIAA0465; OFC4; R74989; trabeculin alpha; trabeculin-alpha; Uncharacterized protein KIAA0754
Common Name MACF1
Gene Symbol MACF1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FSGAALEKEQHLGLLHVRAKDYDTRLDCGYFNTLDSSQVPNAVELIAHVDIMRDTSTVSKEECEKVPFSPRTAEFKSRQPADLDSLEKLDPG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less