missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LUZP4 (aa 208-308) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100048
Description
Highest antigen sequence indentity to the following orthologs: Mouse (30%), Rat (30%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61241 (PA5-61241. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Export adapter involved in mRNA nuclear export in cancer cells. Binds and enhances the RNA-binding activity of the nuclear RNA export factor NXF1. Can restore mRNA export function in cells compromised by loss of mRNA export adapters.Specifications
| Q9P127 | |
| Blocking Assay, Control | |
| 51213 | |
| 100 μL | |
| Cancer/testis antigen 28; CT28; CT-28; CT-8; HOM-TES-85; HOM-TES-85 tumor antigen; leucine zipper protein 4; LUZP4; Tumor antigen HOM-TES-85 | |
| LUZP4 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human LUZP4 (aa 208-308) Control Fragment | |
| RUO | |
| LUZP4 | |
| Unconjugated | |
| Recombinant | |
| SERSHGHSERSHGHSERSHGHSKRSRSQGDLVDTQSDLIATQRDLIATQKDLIATQRDLIATQRDLIVTQRDLVATERDLINQSGRSHGQSERHQRYSTGK | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |