missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human LTA4H ELISA Kit
ELISA
Supplier: Invitrogen™ EH308RB
Description
Human LTA4H quantitates human LTA4H in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human LTA4H.
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.Chemical Identifiers
| Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
| 2.1 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| LTA4H | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| LTA4H | |
| 4 hrs 45 mins |
| 2.1 to 500 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4048 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| Leukotriene A(4) hydrolase; leukotriene A4 hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA4; LTA4 hydrodase; LTA-4 hydrolase; LTA4H; testicular secretory protein Li 27 | |
| 2.1 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| LTA4H | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 2.1 to 500 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 4048 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| LTA4H | |
| 4 hrs 45 mins |