missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LSMEM2 (aa 1-50) Control Fragment Recombinant Protein

Catalog No. RP104027
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104027 Supplier Invitrogen™ Supplier No. RP104027

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140359 (PA5-140359. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LSMEM2 (Leucine Rich Single-Pass Membrane Protein 2) is a Protein Coding gene. Predicted to be integral component of membrane.

Specifications

Accession Number Q8N112
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 132228
Name Human LSMEM2 (aa 1-50) Control Fragment
Quantity 100 μL
Gene Alias C3orf45Leucine-rich single-pass membrane protein 2; LSMEM2
Common Name LSMEM2
Gene Symbol LSMEM2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPSLAPDCPLLAMPEETQEDSVAPMMPSQRSRGPLAPNHVHEVCLHQVES
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less