missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LRRTM3 (aa 302-385) Control Fragment Recombinant Protein

Catalog No. RP109210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109210 Supplier Invitrogen™ Supplier No. RP109210
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139900 (PA5-139900. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Recent evidence suggests that LRRTM3 promotes the processing of amyloid-precursor protein (APP) by the beta-secretase BACE, leading to the proteolytic production of the A-beta42 peptide that is the main component of amyloid plaques. Furthermore, LRRTM3 maps to a region of chromosome 10 linked to both late-onset Alzheimer's disease (AD) and elevated plasma A-beta42 levels, suggesting that LRRTM3 is a functional and positional candidate gene for AD.

Specifications

Accession Number Q86VH5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 347731
Name Human LRRTM3 (aa 302-385) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9630044H04Rik; leucine rich repeat transmembrane neuronal 3; leucine-rich repeat transmembrane neuronal protein 3; LRRTM3; UNQ803/PRO1693
Common Name LRRTM3
Gene Symbol LRRTM3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ISLNDISLAGNIWECSRNICSLVNWLKSFKGLRENTIICASPKELQGVNVIDAVKNYSICGKSTTERFDLARALPKPTFKPKLP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less